2-Amino-3-bromo-5-nitrobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A151557-100G A151557-1G A151557-25G A151557-5G
ALA151557-100GEA 594.61 USD
ALA151557-100G ALA151557-1G ALA151557-25G ALA151557-5G
2-Amino-3-bromo-5-nitrobenzonitrile ≥98%
2-Amino-3-bromo-5-nitrobenzonitrile
Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
Melting Pt: 180-185°C
MDL Number: MFCD00054185
CAS Number: 17601-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bca8547d79f51f2b835908046c0b997b4901562c&vid=bca8547d79f51f2b835908046c0b997b4901562c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR