2-Acetamido-5-bromobenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Acetamido-5-bromobenzoic Acid, N-Acetyl-5-bromoanthranilic acid

A151556-1G A151556-25G A151556-5G
ALA151556-1GEA 27.46 USD
ALA151556-1G ALA151556-25G ALA151556-5G
2-Acetamido-5-bromobenzoic acid ≥98%
2-Acetamido-5-bromobenzoic acid
Formula: C₉H₈BrNO₃
Melting Pt: 218 °C
MDL Number: MFCD00040904
CAS Number: 38985-79-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29243b354b7e3ac3492a1eac28aaa239b0697101&vid=29243b354b7e3ac3492a1eac28aaa239b0697101&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR