3-Amino-3-(4-chlorophenyl)propionic acid ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Amino-3-(4-chlorophenyl)propanoic acid, 3-Amino-3-(4-chlorophenyl)propionic Acid, 3-Amino-3-(p-chlorophenyl)propionic acid

A151555-250MG A151555-25G
ALA151555250MGEA 10.36 USD
ALA151555250MG ALA151555-25G
3-Amino-3-(4-chlorophenyl)propionic acid ≥97% (by HPLC)
3-Amino-3-(4-chlorophenyl)propionic acid
Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Melting Pt: 223°C
MDL Number: MFCD00126370
CAS Number: 19947-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=882fe030a07ce8e82538880c371d6f550cc18eb1&vid=882fe030a07ce8e82538880c371d6f550cc18eb1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR