5-Acetylsalicylic acid ≥98% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Acetyl-2-hydroxybenzoic acid, 5-Acetylsalicylic Acid

A151554-100G A151554-1G A151554-25G A151554-5G
ALA151554-100GEA 766.56 USD
ALA151554-100G ALA151554-1G ALA151554-25G ALA151554-5G
5-Acetylsalicylic acid ≥98% (by HPLC, titration analysis)
5-Acetylsalicylic acid
Formula: C₉H₈O₄
MW: 180.15 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00013978
CAS Number: 13110-96-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=448fdcfd73177fe2935b282172b398f7d0f80a8b&vid=448fdcfd73177fe2935b282172b398f7d0f80a8b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR