[2-(Aminomethyl)phenyl]acetic acid ≥97% (by HPLC, titration analysis)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
[2-(Aminomethyl)phenyl]acetic Acid, 2-Aminomethylphenylacetic acid, 2-(2-(Aminomethyl)phenyl)acetic acid
A151553-100G
A151553-10G
A151553-1G
A151553-250MG
A151553-25G
A151553-5G
ALA151553-100GEA
2342.61
USD
ALA151553-100G
ALA151553-10G
ALA151553-1G
ALA151553250MG
ALA151553-25G
ALA151553-5G
[2-(Aminomethyl)phenyl]acetic acid ≥97% (by HPLC, titration analysis)
[2-(Aminomethyl)phenyl]acetic acid
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038440 CAS Number: 40851-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...