6A-Azido-6A-deoxy-beta-cyclodextrin ≥85% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Mono(6-azido-6-desoxy)-β-cyclodextrin, Mono-6A-azido-β-cyclodextrin, 6-Deoxy-6-azido-β-cyclodextrin, 6-Monoazido-6-monodeoxy-β-cyclodextrin, 6-Azido-6-deoxycyclomaltoheptaose, 6-Monoazido-6-deoxy-β-cyclodextrin, 6-Monodeoxy-6-monoazido-beta-cyclodextrin, 2,4,7,9,12,14,17,19,22,24,27,29,32,34-Tetradecaoxaoctacyclo[31.2.2.23,6.28,11.213,16.218,21.223,26.228,31]nonatetracontane, β-cyclodextrin derivative, 6-Monoazido-β-cyclodextrin, 6A-Azido-6A-deoxy-β-cyclodextrin

A151547-100MG A151547-10MG A151547-25MG
ALA151547100MGEA 359.01 USD
ALA151547100MG ALA151547-10MG ALA151547-25MG
6A-Azido-6A-deoxy-beta-cyclodextrin ≥85% (by HPLC)
6A-Azido-6A-deoxy-beta-cyclodextrin
Formula: C₄₂H₆₉N₃O₃₄
MW: 1160.00 g/mol
Melting Pt: 206°C
Storage Temperature: Freezer
MDL Number: MFCD05864973
CAS Number: 98169-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2cd9bdc5d2ffc5401f1715c63dae687c1cb0146&vid=a2cd9bdc5d2ffc5401f1715c63dae687c1cb0146&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR