Aminomalononitrile-p-toluenesulfonate ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Aminomalononitrile p-Toluenesulphonate, Aminomalononitrile p-Toluenesulphonic acid, Aminomalononitrile p-Toluenesulfonic acid, Dicyanomethylammonium p-Toluenesulphonate, 2-Aminomalononitrile 4-methylbenzenesulphonate, Dicyanomethylammonium p-Toluenesulfonate

A151543-5G A151543-100G A151543-1G A151543-250MG A151543-25G
ALA151543-5GEA 58.81 USD
ALA151543-5G ALA151543-100G ALA151543-1G ALA151543250MG ALA151543-25G
Aminomalononitrile-p-toluenesulfonate ≥98% (by titration)
Aminomalononitrile-p-toluenesulphonate

Application:Aminomalononitrile p-toluenesulfonate was used in the syntheiss of 1-substituted 5-amino-4-cyano-2-hydroxyimidazoles.

Formula: C₃H₃N₃
MW: 253.28 g/mol
Melting Pt: 164 °C
Storage Temperature: Freezer
MDL Number: MFCD00012848
CAS Number: 5098-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b7f1ddcb6ee8e9e1b0f8d859b430ae38114240e&vid=7b7f1ddcb6ee8e9e1b0f8d859b430ae38114240e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR