2-Amino-1,1,3-tricyano-1-propene ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Malononitrile Dimer, 2-Aminoprop-1-ene-1,1,3-tricarbonitrile

A151542-100G A151542-1G A151542-25G A151542-5G
ALA151542-100GEA 152.86 USD
ALA151542-100G ALA151542-1G ALA151542-25G ALA151542-5G
2-Amino-1,1,3-tricyano-1-propene ≥98%
2-Amino-1,1,3-tricyano-1-propene
Formula: C₆H₄N₄
MW: 132.13 g/mol
Melting Pt: 169-172°C
Storage Temperature: Ambient
MDL Number: MFCD00001851
CAS Number: 868-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=592a7c5ef63f56c32d21f3fa6858cfb4c015b602&vid=592a7c5ef63f56c32d21f3fa6858cfb4c015b602&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR