4-Aminodiphenylamine hydrochloride ≥97%
Supplier: Aladdin Scientific
|
Warning
|
A151541-1G
A151541-25G
A151541-5G
ALA151541-1GEA
10.36
USD
ALA151541-1G
ALA151541-25G
ALA151541-5G
4-Aminodiphenylamine hydrochloride ≥97%
4-Aminodiphenylamine hydrochloride
Formula:
C₁₂H₁₂N₂ Melting Pt: 241°C Storage Temperature: Ambient |
MDL Number:
MFCD00012988 CAS Number: 2198-59-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...