Acetohydrazide ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Acetic hydrazide, Acethydrazide

A151537-100G A151537-25G A151537-500G A151537-5G
ALA151537-100GEA 119.61 USD
ALA151537-100G ALA151537-25G ALA151537-500G ALA151537-5G
Acetohydrazide ≥98%
Acetohydrazide

Acethydrazide can be used in the synthesis of (E)-N′-(2-hydroxybenzylidene)acetohydrazide, an ONO pincer ligand.

Formula: C₂H₆N₂O
MW: 74.08 g/mol
Boiling Pt: 134°C
Melting Pt: 57-60°C
Flash Pt: >110°C
Storage Temperature: Ambient
MDL Number: MFCD00007610
CAS Number: 1068-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7853cad6cace4de1c84931a7f51682acead19229&vid=7853cad6cace4de1c84931a7f51682acead19229&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR