4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine ≥95%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A151535-1G A151535-250MG A151535-5G
ALA151535-1GEA 98.71 USD
ALA151535-1G ALA151535250MG ALA151535-5G
4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine ≥95%, Moligand™
4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine
Formula: C₁₁H₉N₅
Melting Pt: 214-217 °C
Storage Temperature: Freezer
MDL Number: MFCD00022901
CAS Number: 5334-30-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb896e5d196a1b080188e8ca8e459d7f2244b7fd&vid=bb896e5d196a1b080188e8ca8e459d7f2244b7fd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR