5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 3-(4-Chlorophenyl)-1-phenyl-1H-pyrazol-5-amine

A151520-1G A151520-200MG A151520-5G
ALA151520-1GEA 100.61 USD
ALA151520-1G ALA151520200MG ALA151520-5G
5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole ≥98%
5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole
Formula: C₁₅H₁₂ClN₃
MW: 269.73 g/mol
Melting Pt: 190.0 to 194.0 °C
MDL Number: MFCD00197059
CAS Number: 19652-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d73028483bedfc532195d7ffe81b0a1fed1c66b&vid=1d73028483bedfc532195d7ffe81b0a1fed1c66b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR