3-(4-Chlorophenyl)-1H-pyrazol-5-amine ≥98% (by HPLC, titration analysis)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
3-Amino-5-(4-chlorophenyl)pyrazole, 5-Amino-3-(4-chlorophenyl)-1H-pyrazole, 5-(4-Chlorophenyl)-1H-pyrazol-3-amine
A151518-1G
A151518-200MG
A151518-5G
ALA151518-1GEA
42.66
USD
ALA151518-1G
ALA151518200MG
ALA151518-5G
3-(4-Chlorophenyl)-1H-pyrazol-5-amine ≥98% (by HPLC, titration analysis)
3-(4-Chlorophenyl)-1H-pyrazol-5-amine
Formula:
C₉H₈ClN₃ Melting Pt: 175 °C |
MDL Number:
MFCD00053046 CAS Number: 78583-81-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...