3-Amino-5-methyl-1H-pyrazole ≥96% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151511-100G A151511-25G A151511-5G
ALA151511-100GEA 188.01 USD
ALA151511-100G ALA151511-25G ALA151511-5G
3-Amino-5-methyl-1H-pyrazole ≥96% (by GC)
3-Amino-5-methyl-1H-pyrazole

Complexation of the amino- and carboxyl-protected tripeptide with 3-amino-5-methylpyrazole was studied by low-temperature NMR experiments in a freonic solvent.application:3-Amino-5-methylpyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It was also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones.

Formula: C₄H₇N₃
Melting Pt: 41 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075180
CAS Number: 31230-17-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5caf08f52009303f7a4c723bc73ff65ab9c7e01&vid=c5caf08f52009303f7a4c723bc73ff65ab9c7e01&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR