5-Amino-3-methyl-1-p-tolylpyrazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151510-1G A151510-250MG
ALA151510-1GEA 22.71 USD
ALA151510-1G ALA151510250MG
5-Amino-3-methyl-1-p-tolylpyrazole ≥97%
5-Amino-3-methyl-1-p-tolylpyrazole
Formula: C₁₁H₁₃N₃
Melting Pt: 118 °C
MDL Number: MFCD00020725
CAS Number: 62535-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43ed5afb0b8f5a78b7c79e73816fbb12c48a56a5&vid=43ed5afb0b8f5a78b7c79e73816fbb12c48a56a5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR