5-Amino-1-phenyl-1H-pyrazole ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151509-1G A151509-25G A151509-5G
ALA151509-1GEA 11.31 USD
ALA151509-1G ALA151509-25G ALA151509-5G
5-Amino-1-phenyl-1H-pyrazole ≥98%
5-Amino-1-phenyl-1H-pyrazole
Formula: C₉H₉N₃
Melting Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00020734
CAS Number: 826-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ec24d0760a9e95d1a7ec8e5a0dbde5ea8c4af3c&vid=5ec24d0760a9e95d1a7ec8e5a0dbde5ea8c4af3c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR