5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A151507-1G A151507-200MG
ALA151507-1GEA 41.71 USD
ALA151507-1G ALA151507200MG
5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole ≥96%
5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole
Formula: C₁₀H₁₀BrN₃
Melting Pt: 103-107°C
MDL Number: MFCD04039926
CAS Number: 69464-98-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18a08cfe20dd55c85ae4ad3102e0ab9947b21502&vid=18a08cfe20dd55c85ae4ad3102e0ab9947b21502&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR