(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate ≥97%
Supplier: ALADDIN SCIENTIFIC
A151506-100MG
A151506-1G
A151506-250MG
A151506-25MG
ALA151506100MGEA
289.66
USD
ALA151506100MG
ALA151506-1G
ALA151506250MG
ALA151506-25MG
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate ≥97%
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate
Formula:
C₂₁H₂₂ClN₃O Melting Pt: 219 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD10566990 CAS Number: 919102-70-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...