3-Aminoisonicotinic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Aminoisonicotinic Acid, 3-Aminopyridine-4-carboxylic acid

A151502-100G A151502-10G A151502-1G A151502-25G A151502-5G
ALA151502-100GEA 1576.91 USD
ALA151502-100G ALA151502-10G ALA151502-1G ALA151502-25G ALA151502-5G
3-Aminoisonicotinic acid ≥98% (by HPLC)
3-Aminoisonicotinic acid
Formula: C₆H₆N₂O₂
MW: 138.13
Melting Pt: 305-310 °C
MDL Number: MFCD00137840
CAS Number: 7579-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e7aea45111445d4feefb2fd5ae5dc7ccb77d8ea&vid=3e7aea45111445d4feefb2fd5ae5dc7ccb77d8ea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR