5-Acetylindan ≥95%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 1-(2,3-Dihydro-1H-inden-5-yl)ethan-1-one, 1-(2,3-Dihydro-1H-inden-5-yl)ethanone

A151500-1G A151500-25G A151500-50G A151500-5G
ALA151500-1GEA 25.56 USD
ALA151500-1G ALA151500-25G ALA151500-50G ALA151500-5G
5-Acetylindan ≥95%
5-Acetylindan
Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Boiling Pt: 85°C
MDL Number: MFCD00021246
CAS Number: 4228-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR