3-Aminobenzophenone ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: m-Aminobenzophenone

A151498-1G A151498-25G A151498-5G
ALA151498-1GEA 10.36 USD
ALA151498-1G ALA151498-25G ALA151498-5G
3-Aminobenzophenone ≥98%
3-Aminobenzophenone

Product Introduction:3-Aminobenzophenone forms cyclodextrin (α and β) based nanostructures through the supramolecular self assembly.  Application3-Aminobenzophenone has been used in the synthesis of racemic benzophenone ureas, chiral photoaffinity labeling probes.

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Melting Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00017103
CAS Number: 2835-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23523206e005f9df14a3123fb6bbb65d4a83391a&vid=23523206e005f9df14a3123fb6bbb65d4a83391a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR