4-Aminobenzophenone ≥98% (by titration)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).
Formula:
C₁₃H₁₁NO Boiling Pt: 245-247°C Melting Pt: 121-124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007895 CAS Number: 1137-41-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...