Allyl(tert-butyl)dimethylsilane ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A151492-1G A151492-25G A151492-5G
ALA151492-1GEA 52.16 USD
ALA151492-1G ALA151492-25G ALA151492-5G
Allyl(tert-butyl)dimethylsilane ≥96%
Allyl(tert-butyl)dimethylsilane
Formula: C₉H₂₀Si
Boiling Pt: 152°C
Density: 0.77 g/cm³
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD18207712
CAS Number: 74472-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3584261b2058cf90f76e23cb21455a7f314551f7&vid=3584261b2058cf90f76e23cb21455a7f314551f7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR