4-Amino-4'-nitrobiphenyl ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151491-1G A151491-250MG A151491-5G
ALA151491-1GEA 177.56 USD
ALA151491-1G ALA151491250MG ALA151491-5G
4-Amino-4'-nitrobiphenyl ≥95%
4-Amino-4'-nitrobiphenyl
Formula: C₁₂H₁₀N₂O₂
Melting Pt: 204 °C
MDL Number: MFCD00093527
CAS Number: 1211-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203ef38418667c163908d4f95912904f9ff8c2f4&vid=203ef38418667c163908d4f95912904f9ff8c2f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR