ɑ,ε-Dibenzoyl-DL-lysine ≥99%
Supplier: ALADDIN SCIENTIFIC
A151490-1G
A151490-5G
ALA151490-1GEA
76.86
USD
ALA151490-1G
ALA151490-5G
ɑ,ε-Dibenzoyl-DL-lysine ≥99%
alpha,epsilon-Dibenzoyl-DL-lysine
Formula:
C₂₀H₂₂N₂O₄ Melting Pt: 146 °C |
MDL Number:
MFCD00059684 CAS Number: 32513-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...