Ammonium 8-anilinonaphthalene-1-sulphonate ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Ammonium 8-(phenylamino)naphthalene-1-sulphonate, 8-Anilino-1-naphthalenesulphonic acid Ammonium salt, Ammonium 8-Anilino-1-naphthalenesulphonate, 8-Anilinonaphthalene-1-sulphonic acid ammonium salt, ANS-NH4

A151487-100G A151487-1G A151487-25G A151487-5G
ALA151487-100GEA 1710.86 USD
ALA151487-100G ALA151487-1G ALA151487-25G ALA151487-5G
Ammonium 8-anilinonaphthalene-1-sulphonate ≥95%
Ammonium 8-anilinonaphthalene-1-sulphonate
Formula: C₁₆H₁₆N₂O₃S
Melting Pt: 237 °C
Storage Temperature: Ambient
MDL Number: MFCD00012560
CAS Number: 28836-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7a97dba8deb5d1a124d8e6813bb57770691fa06&vid=c7a97dba8deb5d1a124d8e6813bb57770691fa06&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR