ATBTA-Eu3+ ≥90% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION

A151484-1MG A151484-5MG
ALA151484-1MGEA 20.81 USD
ALA151484-1MG ALA151484-5MG
ATBTA-Eu3+ ≥90% (by HPLC, titration analysis)
ATBTA-Eu3+
Formula: C₃₇H₃₀EuN₆NaO₈
MW: 861.635 g/mol
Storage Temperature: Ambient
CAS Number: 601494-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a1cc41cb2921e735da6a78aa0304de3b1cae572&vid=3a1cc41cb2921e735da6a78aa0304de3b1cae572&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR