N-Acetonylpyridinium chloride ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151480-5G A151480-1G A151480-25G
ALA151480-5GEA 45.51 USD
ALA151480-5G ALA151480-1G ALA151480-25G
N-Acetonylpyridinium chloride ≥97% (by HPLC)
N-Acetonylpyridinium chloride
Formula: C₈H₁₀ClNO
Melting Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00031949
CAS Number: 42508-60-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86787cd79d343bd5ff5e490c4964907cc00d3d75&vid=86787cd79d343bd5ff5e490c4964907cc00d3d75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR