11-Azido-3,6,9-trioxaundecanoic acid ≥97% (by titration)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
Azido-PEG3-carboxylate, 2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]acetic acid, 11-Azido-3,6,9-trioxaundecanoic Acid, Azido-PEG3-acetic acid
A151476-1G
A151476-200MG
ALA151476-1GEA
344.76
USD
ALA151476-1G
ALA151476200MG
11-Azido-3,6,9-trioxaundecanoic acid ≥97% (by titration)
11-Azido-3,6,9-trioxaundecanoic acid
Formula:
C₈H₁₅N₃O₅ MW: 233.22 g/mol |
CAS Number:
172531-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...