1-Amino-1-cyclobutanecarboxylic acid hydrochloride ≥98% (by titration)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
1-Aminocyclobutanecarboxylic acid hydrochloride, 1-Aminocyclobutanecarboxylic Acid HCl
A151474-1G
A151474-25G
A151474-5G
ALA151474-1GEA
33.16
USD
ALA151474-1G
ALA151474-25G
ALA151474-5G
1-Amino-1-cyclobutanecarboxylic acid hydrochloride ≥98% (by titration)
1-Amino-1-cyclobutanecarboxylic acid hydrochloride
Formula:
C₅H₁₀ClNO₂ MW: 151.59 g/mol Melting Pt: 226°C |
MDL Number:
MFCD08752285 CAS Number: 98071-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...