1-Aminocyclopentanecarboxylic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 1-Aminocyclopentanecarboxylic Acid, Cycloleucine, 1-Aminocyclopentane-1-carboxylic acid

A151472-1G A151472-250G A151472-50G A151472-5G
ALA151472-1GEA 22.71 USD
ALA151472-1G ALA151472-250G ALA151472-50G ALA151472-5G
1-Aminocyclopentanecarboxylic acid ≥98%
1-Aminocyclopentanecarboxylic acid
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 330°C
MDL Number: MFCD00001381
CAS Number: 52-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5193c8436cce2720f7f688c5b2b156bb5c738fbd&vid=5193c8436cce2720f7f688c5b2b156bb5c738fbd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR