6-Amino-2-naphthoic acid ≥97% (by HPLC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
6-Amino-2-naphthoic acid, 6-Amino-2-naphthalenecarboxylic acid, 6-Amino-2-naphthoic Acid
A151467-1G
A151467-250MG
A151467-50MG
A151467-5G
ALA151467-1GEA
36.01
USD
ALA151467-1G
ALA151467250MG
ALA151467-50MG
ALA151467-5G
6-Amino-2-naphthoic acid ≥97% (by HPLC)
6-Amino-2-naphthoic acid
Formula:
C₁₁H₉NO₂ MW: 187.1947 g/mol Melting Pt: 225°C Storage Temperature: Ambient |
MDL Number:
MFCD01861831 CAS Number: 116668-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...