4-Amino-3-nitrobenzoic acid ≥95% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151466-100G
A151466-250G
A151466-25G
A151466-5G
ALA151466-100GEA
126.26
USD
ALA151466-100G
ALA151466-250G
ALA151466-25G
ALA151466-5G
4-Amino-3-nitrobenzoic acid ≥95% (by HPLC)
4-Amino-3-nitrobenzoic acid
Formula:
C₇H₆N₂O₄ Melting Pt: 280 °C |
MDL Number:
MFCD00017009 CAS Number: 1588-83-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...