ɑ-Methyllevulinic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: α-Methyllevulinic acid, 2-Methyl-4-oxopentanoic acid, 2-Methyl-4-oxopentanoic Acid

A151464-1G A151464-250MG
ALA151464-1GEA 317.21 USD
ALA151464-1G ALA151464250MG
ɑ-Methyllevulinic acid ≥98%
alpha-Methyllevulinic acid
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 165°C
Density: 1.09 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00143584
CAS Number: 6641-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=379557f14367e1837ca89b6d3379188b2f36f35b&vid=379557f14367e1837ca89b6d3379188b2f36f35b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR