4-Amino-5-chloro-2-methoxybenzoic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Metoclopramide Impurity C, 4-Amino-5-chloro-o-anisic acid

A151461-1G A151461-25G A151461-5G A151461-100G
ALA151461-1GEA 9.41 USD
ALA151461-1G ALA151461-25G ALA151461-5G ALA151461-100G
4-Amino-5-chloro-2-methoxybenzoic acid ≥98% (by HPLC)
4-Amino-5-chloro-2-methoxybenzoic acid
Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
Melting Pt: 205-208°C
MDL Number: MFCD00038794
CAS Number: 7206-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7843d03f05dfb9e6490dd308e89d4ab25c91f2d3&vid=7843d03f05dfb9e6490dd308e89d4ab25c91f2d3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR