5-Amino-2-nitrobenzoic acid ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Nitro-5-aminobenzoic acid, NSC 74455, 5-Amino-2-nitrobenzoic Acid, 3-Carboxy-4-nitroaniline
A151458-1G
A151458-250MG
A151458-25G
A151458-5G
ALA151458-1GEA
16.06
USD
ALA151458-1G
ALA151458250MG
ALA151458-25G
ALA151458-5G
5-Amino-2-nitrobenzoic acid ≥98% (by HPLC)
5-Amino-2-nitrobenzoic acid
Reactant for: . Two-component dendritic chain reactions . Enzymic activation of hydrophobic self-immolative dendrimers . Preparation of insulin receptor tyrosine kinase activator . Preparation of polymer-bound diazonium salts using Merrifield resin-bound piperazine
Formula:
C₇H₆N₂O₄ MW: 182.14 g/mol Boiling Pt: 481 Melting Pt: 234 |
MDL Number:
MFCD00035748 CAS Number: 13280-60-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...