3-Amino-4-chlorobenzoic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151457-100G A151457-25G A151457-5G
ALA151457-100GEA 78.76 USD
ALA151457-100G ALA151457-25G ALA151457-5G
3-Amino-4-chlorobenzoic acid ≥98% (by HPLC)
3-Amino-4-chlorobenzoic acid
Formula: C₇H₆ClNO₂
Melting Pt: 216 °C
MDL Number: MFCD00007671
CAS Number: 2840-28-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3fb9a5b28c86b1c45d3fa3f9bd850d67ffc8862&vid=a3fb9a5b28c86b1c45d3fa3f9bd850d67ffc8862&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR