3-Aminosalicylic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151456-1G A151456-250MG A151456-5G
ALA151456-1GEA 86.36 USD
ALA151456-1G ALA151456250MG ALA151456-5G
3-Aminosalicylic acid ≥98% (by HPLC)
3-Aminosalicylic acid

3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.

Formula: C₇H₇NO₃
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00010299
CAS Number: 570-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b14d282f51c02eb2ec02a2d025a32bc1edcc41f4&vid=b14d282f51c02eb2ec02a2d025a32bc1edcc41f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR