3-Acetylpyridine N-Oxide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151453-1G A151453-200MG
ALA151453-1GEA 109.16 USD
ALA151453-1G ALA151453200MG
3-Acetylpyridine N-Oxide ≥98%
3-Acetylpyridine N-Oxide
Formula: C₇H₇NO₂
Melting Pt: 146 °C
MDL Number: MFCD00955798
CAS Number: 14188-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5b09852aa725046e01270278d63797ef732d07&vid=4d5b09852aa725046e01270278d63797ef732d07&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR