Pentyl formate ≥95% (by GC)

Supplier: Aladdin Scientific
Warning

A151451-5ML A151451-100ML A151451-25ML
ALA151451-5MLEA 9.41 USD
ALA151451-5ML ALA151451100ML ALA151451-25ML
Pentyl formate ≥95% (by GC)
Pentyl formate
Formula: C₆H₁₂O₂
Boiling Pt: 130°C
Density: 0.89 g/cm³
Flash Pt: 32°C
MDL Number: MFCD00040440
CAS Number: 638-49-3
UN: 1109
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4f49dabd559ad0e30be5ce6eb3b6b66289bdd3a&vid=e4f49dabd559ad0e30be5ce6eb3b6b66289bdd3a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR