3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid ≥98% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3'-Amino-2'-hydroxybiphenyl-3-carboxylic Acid, 3'-Amino-2'-hydroxy-[1,1'-biphenyl]-3-carboxylic acid, 3-(3-Amino-2-hydroxyphenyl)benzoic acid

A151448-10G A151448-1G A151448-250MG A151448-5G
ALA151448-10GEA 1652.91 USD
ALA151448-10G ALA151448-1G ALA151448250MG ALA151448-5G
3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid ≥98% (by HPLC, titration analysis)
3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid
Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
Melting Pt: 222 °C
MDL Number: MFCD10001467
CAS Number: 376592-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2404486cb617c2774a928774fb84f1683908b361&vid=2404486cb617c2774a928774fb84f1683908b361&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR