3-Aminofluoranthene 95

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Fluoranthen-3-amine

A151447-100MG A151447-1G A151447-250MG A151447-5G
ALA151447100MGEA 9.41 USD
ALA151447100MG ALA151447-1G ALA151447250MG ALA151447-5G
3-Aminofluoranthene 95
3-Aminofluoranthene
Formula: C₁₆H₁₁N
MW: 217.2700 g/mol
Melting Pt: 117-121°C
MDL Number: MFCD00001185
CAS Number: 2693-46-1
EINECS: 220-263-7
UN: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=290de00061020533a0940406827cf32dd0d67b56&vid=290de00061020533a0940406827cf32dd0d67b56&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR