1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151446-25G A151446-5G
ALA151446-25GEA 69.26 USD
ALA151446-25G ALA151446-5G
1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride ≥98% (by HPLC)
1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride
Formula: C₁₅H₂₃NO₂
Melting Pt: 176 °C
MDL Number: MFCD06658143
CAS Number: 130198-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d51eaed652c9171718c05af8bac49c9b246ff55&vid=6d51eaed652c9171718c05af8bac49c9b246ff55&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR