4-Amino-3-nitrobenzophenone ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151440-25G
A151440-5G
ALA151440-25GEA
72.11
USD
ALA151440-25G
ALA151440-5G
4-Amino-3-nitrobenzophenone ≥98%
4-Amino-3-nitrobenzophenone
Formula:
C₁₃H₁₀N₂O₃ Melting Pt: 141°C |
MDL Number:
MFCD00007154 CAS Number: 31431-19-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...