2-Amino-4'-chlorobenzophenone ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (2-Aminophenyl)(4-chlorophenyl)methanone

A151438-1G A151438-25G A151438-5G
ALA151438-1GEA 13.21 USD
ALA151438-1G ALA151438-25G ALA151438-5G
2-Amino-4'-chlorobenzophenone ≥98%
2-Amino-4'-chlorobenzophenone
Formula: C₁₃H₁₀ClNO
Melting Pt: 101-105°C
Storage Temperature: Ambient
MDL Number: MFCD00025193
CAS Number: 2894-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e2eba0d8d62589271e44a7bea4ee63feb37f31d&vid=9e2eba0d8d62589271e44a7bea4ee63feb37f31d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR