1-Aminopyrene ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Pyren-1-ylamine, Pyren-1-amine

A151433-5G A151433-1G A151433-250MG A151433-25G
ALA151433-5GEA 188.01 USD
ALA151433-5G ALA151433-1G ALA151433250MG ALA151433-25G
1-Aminopyrene ≥98% (by HPLC)
1-Aminopyrene
Formula: C₁₆H₁₁N
MW: 217.27 g/mol
Melting Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00004140
CAS Number: 1606-67-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fec3ddd2e95155df7c46144e10da589d6187d88b&vid=fec3ddd2e95155df7c46144e10da589d6187d88b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR