2-Amino-3-hydroxyanthraquinone ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

A151431-1G A151431-25G A151431-5G
ALA151431-1GEA 9.41 USD
ALA151431-1G ALA151431-25G ALA151431-5G
2-Amino-3-hydroxyanthraquinone ≥98%
2-Amino-3-hydroxyanthraquinone
Formula: C₁₄H₉NO₃
MW: 239.23 g/mol
MDL Number: MFCD00059499
CAS Number: 117-77-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f70d8dd1f1cf661d058bbb1af89f4f0ce81b2260&vid=f70d8dd1f1cf661d058bbb1af89f4f0ce81b2260&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR