4-Amino-4'-(N,N-dimethylamino)stilbene ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N,N-Dimethyl-4,4'-stilbenediamine, 4-(4-Aminostyryl)-N,N-dimethylaniline

A151429-1G A151429-250MG
ALA151429-1GEA 169.96 USD
ALA151429-1G ALA151429250MG
4-Amino-4'-(N,N-dimethylamino)stilbene ≥98%
4-Amino-4'-(N,N-dimethylamino)stilbene
Formula: C₁₆H₁₈N₂
MW: 238.33 g/mol
Melting Pt: 172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059189
CAS Number: 22525-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=304c5e31beba7473a78c456341749914f99eee5e&vid=304c5e31beba7473a78c456341749914f99eee5e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR