2-Amino-N-methylbenzamide ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A151423-100G A151423-1G A151423-25G A151423-5G
ALA151423-100GEA 3188.11 USD
ALA151423-100G ALA151423-1G ALA151423-25G ALA151423-5G
2-Amino-N-methylbenzamide ≥98%
2-Amino-N-methylbenzamide
Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00060602
CAS Number: 4141-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR