4-Amino-N-methylbenzamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151421-100G A151421-1G A151421-25G A151421-5G
ALA151421-100GEA 63.56 USD
ALA151421-100G ALA151421-1G ALA151421-25G ALA151421-5G
4-Amino-N-methylbenzamide ≥98%
4-Amino-N-methylbenzamide
Formula: C₈H₁₀N₂O
Melting Pt: 181 °C
MDL Number: MFCD00665792
CAS Number: 6274-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec7e4856f249f009b04151e4ef9641969fd5939d&vid=ec7e4856f249f009b04151e4ef9641969fd5939d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR